COMH580D8246 Combi-Blocks, Inc.

4-(3-Aminophenyl)-2-methylpyrimidine

  • MDL number MFCD00067950
  • Cas Number : 175201-90-8
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9669

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9669
Availability details for
Quantity
  • Availability
  • Add to Cart