COMH580D8247 Combi-Blocks, Inc.

Isoquinoline-3-carboxamide

  • MDL number MFCD00160690
  • Cas Number : 50458-77-0
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1119

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-1119
Availability details for
Quantity
  • Availability
  • Add to Cart