COMH580D8248 Combi-Blocks, Inc.

2,2'-(1,3-Phenylene)diethanol

  • MDL number MFCD09926175
  • Cas Number : 46133-05-5
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2090
Availability details for
Quantity
  • Availability
  • Add to Cart