COMH580D8249 Combi-Blocks, Inc.

tert-Butyl pyridazin-3-ylcarbamate

  • MDL number MFCD24466705
  • Cas Number : 147362-90-1
  • Molecular Weight : 195.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2459

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QJ-2459
Availability details for
Quantity
  • Availability
  • Add to Cart