COMH580D824C Combi-Blocks, Inc.

7-Chloro-2-(4-fluorophenyl)-2h-pyrazolo[3,4-d]pyridazine

  • MDL number MFCD25542221
  • Cas Number : 1956376-33-2
  • Molecular Weight : 248.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7306

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QK-7306
Availability details for
Quantity
  • Availability
  • Add to Cart