COMH580D824D Combi-Blocks, Inc.

Ethyl furo[2,3-b]pyridine-2-carboxylate

  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4584

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4584
Availability details for
Quantity
  • Availability
  • Add to Cart