COMH580D8250 Combi-Blocks, Inc.

2-(4-iodophenyl)-N-isopropylacetamide

  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4826
Availability details for
Quantity
  • Availability
  • Add to Cart