COMH580D8251 Combi-Blocks, Inc.

2-(4-Iodophenyl)-1-(pyrrolidin-1-yl)ethanone

  • Cas Number : 2097676-72-5
  • Molecular Weight : 315.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4827
Availability details for
Quantity
  • Availability
  • Add to Cart