COMH580D8252 Combi-Blocks, Inc.

2-(4-Iodophenyl)-1-(morpholin-4-yl)ethanone

  • Cas Number : 364793-89-5
  • Molecular Weight : 331.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4828
Availability details for
Quantity
  • Availability
  • Add to Cart