COMH580D8253 Combi-Blocks, Inc.

2-(4-Iodophenyl)-1-(piperidin-1-yl)ethanone

  • Cas Number : 1093169-72-2
  • Molecular Weight : 329.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4829
Availability details for
Quantity
  • Availability
  • Add to Cart