COMH580D8256 Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)acetamide

  • Cas Number : 2377920-30-2
  • Molecular Weight : 275.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4857
Availability details for
Quantity
  • Availability
  • Add to Cart