COMH580D8257 Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-N-methylacetamide

  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4858

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4858
Availability details for
Quantity
  • Availability
  • Add to Cart