COMH580D8258 Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-N-ethylacetamide

  • Cas Number : 2094647-74-0
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4859

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4859
Availability details for
Quantity
  • Availability
  • Add to Cart