COMH580D825A Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-N-cyclopropylacetamide

  • Molecular Weight : 315.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4861

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4861
Availability details for
Quantity
  • Availability
  • Add to Cart