COMH580D825B Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-N-propylacetamide

  • Molecular Weight : 317.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4862

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4862
Availability details for
Quantity
  • Availability
  • Add to Cart