COMH580D825C Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-1-(pyrrolidin-1-yl)ethanone

  • Cas Number : 2377920-23-3
  • Molecular Weight : 329.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4863

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4863
Availability details for
Quantity
  • Availability
  • Add to Cart