COMH580D825D Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-1-(morpholin-4-yl)ethanone

  • Molecular Weight : 345.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4864

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4864
Availability details for
Quantity
  • Availability
  • Add to Cart