COMH580D825F Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-1-(piperidin-1-yl)ethanone

  • Cas Number : 2377920-31-3
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4866
Availability details for
Quantity
  • Availability
  • Add to Cart