COMH580D8260 Combi-Blocks, Inc.

2-(2-Bromo-4-nitrophenoxy)-N,N-dimethylacetamide

  • Cas Number : 2377920-28-8
  • Molecular Weight : 303.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4867

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4867
Availability details for
Quantity
  • Availability
  • Add to Cart