COMH580D8263 Combi-Blocks, Inc.

2-Bromo-4-nitrophenoxyacetic acid

  • MDL number MFCD28101250
  • Cas Number : 25300-00-9
  • Molecular Weight : 276.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?AN-6387
Availability details for
Quantity
  • Availability
  • Add to Cart