COMH580D8272 Combi-Blocks, Inc.

2,5-Dimethylpyridin-3-amine

  • MDL number MFCD13175353
  • Cas Number : 89943-02-2
  • Molecular Weight : 122.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-7076
Availability details for
Quantity
  • Availability
  • Add to Cart