COMH580D8274 Combi-Blocks, Inc.

Ethyl endo-3-boc-3-azabicyclo-[3.1.0]hexane-6-carboxylate

  • MDL number MFCD27986920
  • Cas Number : 827599-20-2
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QM-8923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QM-8923
Availability details for
Quantity
  • Availability
  • Add to Cart