COMH580D8279 Combi-Blocks, Inc.

Aberchrome 670

  • MDL number MFCD07368579
  • Cas Number : 94856-25-4
  • Molecular Weight : 352.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5189

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QN-5189
Availability details for
Quantity
  • Availability
  • Add to Cart