COMH580D827F Combi-Blocks, Inc.

4-[(diethylamino)methyl]benzaldehyde

  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6451
Availability details for
Quantity
  • Availability
  • Add to Cart