COMH580D8282 Combi-Blocks, Inc.

1-Bromo-3-(3-chloropropyl)benzene

  • MDL number MFCD09744297
  • Cas Number : 1057673-67-2
  • Molecular Weight : 233.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7356

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7356
Availability details for
Quantity
  • Availability
  • Add to Cart