COMH580D8283 Combi-Blocks, Inc.

Ethyl 2-(5-bromo-3-fluoropyridin-2-yl)acetate

  • MDL number MFCD28740664
  • Cas Number : 1620318-88-8
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7451
Availability details for
Quantity
  • Availability
  • Add to Cart