COMH580D8287 Combi-Blocks, Inc.

Methyl 5-chloronaphthalene-1-carboxylate

  • MDL number MFCD13704868
  • Cas Number : 16650-65-0
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7514
Availability details for
Quantity
  • Availability
  • Add to Cart