COMH580D8288 Combi-Blocks, Inc.

(2E)-3-(3,5-dimethylphenyl)prop-2-enoic acid

  • MDL number MFCD03002760
  • Cas Number : 67688-87-3
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-7517
Availability details for
Quantity
  • Availability
  • Add to Cart