COMH580D82B9 Combi-Blocks, Inc.

4-Amino-5-(2-methylphenyl)-4h-1,2,4-triazole-3-thiol

  • MDL number MFCD04331378
  • Cas Number : 87239-95-0
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4648

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?ST-4648
Availability details for
Quantity
  • Availability
  • Add to Cart