COMH6E40A3BE Combi-Blocks, Inc.

tert-Butyl 2-(tetramethyl-1,3,2-dioxaborolan-2-yl)acetate

  • MDL number MFCD27936366
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1509

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-1509
Availability details for
Quantity
  • Availability
  • Add to Cart