COMH6E40A4DD Combi-Blocks, Inc.

2,2-Dimethyl-4,7-dihydro-1,3-dioxepine

  • MDL number MFCD22054122
  • Cas Number : 1003-83-4
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9150

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QI-9150
Availability details for
Quantity
  • Availability
  • Add to Cart