COMH6E40A51F Combi-Blocks, Inc.

2-(4-Iodophenyl)acetamide

  • Cas Number : 84863-81-0
  • Molecular Weight : 261.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-iodophenyl)acetamide
Inchi Code 1S/C8H8INO/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4H,5H2,(H2,10,11)
Inchi Key RKJMVTJNBPMLGH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QL-4830
Availability details for
Quantity
  • Availability
  • Add to Cart