COMH93D5B61C Combi-Blocks, Inc.

2,4,6-Trimethylphenylboronic acid neopentyl glycol ester

  • MDL number MFCD01318178
  • Cas Number : 214360-78-8
  • Molecular Weight : 232.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8329

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?BB-8329
Availability details for
Quantity
  • Availability
  • Add to Cart