COMH93D5B792 Combi-Blocks, Inc.

1-(2-Methoxybenzoyl)piperazine

  • MDL number MFCD02256018
  • Cas Number : 436099-85-3
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0315
Availability details for
Quantity
  • Availability
  • Add to Cart