COMH93D5B7C0 Combi-Blocks, Inc.

Lidorestat

  • MDL number MFCD08067733
  • Cas Number : 245116-90-9
  • Molecular Weight : 376.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0385

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0385
Availability details for
Quantity
  • Availability
  • Add to Cart