COMH93D5B7C6 Combi-Blocks, Inc.

3-(2-Aminoethyl)-5-methoxy-1h-indole-2-carboxylic acid

  • MDL number MFCD00450677
  • Cas Number : 52648-13-2
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0395

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0395
Availability details for
Quantity
  • Availability
  • Add to Cart