COMH93D5B7F3 Combi-Blocks, Inc.

1-(4-Fluorophenoxy)-3-nitrobenzene

  • MDL number MFCD02246891
  • Cas Number : 263349-49-1
  • Molecular Weight : 233.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0458

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0458
Availability details for
Quantity
  • Availability
  • Add to Cart