COMH93D5B7F5 Combi-Blocks, Inc.

(S)-Ethyl 3-amino-4,4,4-trifluorobutanoate hcl

  • MDL number MFCD07784300
  • Cas Number : 1212216-61-9
  • Molecular Weight : 221.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0460

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0460
Availability details for
Quantity
  • Availability
  • Add to Cart