COMH93D5B834 Combi-Blocks, Inc.

(2R,3R)-3-((tert-Butoxycarbonyl)amino)-2-hydroxy-3-(3-methoxyphenyl)propanoic acid

  • MDL number MFCD07363641
  • Cas Number : 1217672-32-6
  • Molecular Weight : 311.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0566

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0566
Availability details for
Quantity
  • Availability
  • Add to Cart