COMH93D5B863 Combi-Blocks, Inc.

3-Bromo-n-[2-(dimethylamino)ethyl]benzamide

  • MDL number MFCD02916801
  • Cas Number : 425618-78-6
  • Molecular Weight : 271.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0638

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0638
Availability details for
Quantity
  • Availability
  • Add to Cart