COMH93D5B873 Combi-Blocks, Inc.

(R)-Citalopram oxalate

  • MDL number MFCD06407825
  • Cas Number : 219861-53-7
  • Molecular Weight : 414.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0662
Availability details for
Quantity
  • Availability
  • Add to Cart