COMH93D5B89A Combi-Blocks, Inc.

X-Alpha-d-fuc

  • MDL number MFCD04972044
  • Molecular Weight : 392.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0721

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0721
Availability details for
Quantity
  • Availability
  • Add to Cart