COMH93D5B90D Combi-Blocks, Inc.

2,3-Dimethylphenazine

  • MDL number MFCD01861444
  • Cas Number : 6479-88-5
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0903

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-0903
Availability details for
Quantity
  • Availability
  • Add to Cart