COMH93D5B974 Combi-Blocks, Inc.

Di(1h-pyrrol-2-yl)methanone

  • MDL number MFCD01686800
  • Cas Number : 15770-21-5
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1060

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1060
Availability details for
Quantity
  • Availability
  • Add to Cart