COMH93D5B9A2 Combi-Blocks, Inc.

Dimethyl 1-benzyl-1h-imidazole-4,5-dicarboxylate

  • MDL number MFCD00428974
  • Cas Number : 321970-25-6
  • Molecular Weight : 274.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1137
Availability details for
Quantity
  • Availability
  • Add to Cart