COMH93D5B9E3 Combi-Blocks, Inc.

1-(3,4,5-Trimethoxyphenyl)-piperazine hydrochloride

  • MDL number MFCD00017647
  • Cas Number : 116290-71-2
  • Molecular Weight : 288.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1232
Availability details for
Quantity
  • Availability
  • Add to Cart