COMH93D5B9EA Combi-Blocks, Inc.

4-Hydroxy-4'-methoxydihydrochalcone

  • MDL number MFCD00075981
  • Cas Number : 206194-36-7
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1247
Availability details for
Quantity
  • Availability
  • Add to Cart