COMH93D5B9ED Combi-Blocks, Inc.

4-Methyl-1-hydrazino-phthalazine

  • MDL number MFCD01326534
  • Cas Number : 29902-28-1
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1250
Availability details for
Quantity
  • Availability
  • Add to Cart