COMH93D5BA6D Combi-Blocks, Inc.

Oxomemazine

  • MDL number MFCD00868197
  • Cas Number : 3689-50-7
  • Molecular Weight : 330.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1447
Availability details for
Quantity
  • Availability
  • Add to Cart