COMH93D5BA79 Combi-Blocks, Inc.

Artemisinic acid

  • MDL number MFCD00238540
  • Cas Number : 80286-58-4
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1470

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-1470
Availability details for
Quantity
  • Availability
  • Add to Cart